Learn More
Cayman Chemical 5Z8ZTetradecadIenoIcAc50ug
SDP
Catalog No. 502919278
Encompass_Preferred
An internal standard for the quantification of 5(Z),8(Z)-tetradecadienoic acid by GC- or LC-MS
Catalog No. 50-291-9278 Supplier Cayman Chemical Supplier No. 3971150 UG
An internal standard for the quantification of 5(Z),8(Z)-tetradecadienoic acid by GC- or LC-MS