Learn More
Doccol Corporation 6-0 FINE MCAO SUTURE L56RE PK5
SDP
Catalog No. NC1861603
Encompass
Silicone rubber-coated monofilament for MCAO model. Filament size 6-0, diameter 0.09-0.11 mm, length 20mm; diameter with coating 0.21 +/- 0.02 mm; coating length 5-6 mm, suitable for MCAO with reperfusion, or permanent MCAO. Reusable. Pack of 5 filaments.
This item is not returnable. View return policy
Catalog No. NC1861603 Supplier Doccol Corporation Supplier No. 602156PK5RE
Silicone rubber-coated monofilament for MCAO model. Filament size 6-0, diameter 0.09-0.11 mm, length 20mm; diameter with coating 0.21 +/- 0.02 mm; coating length 5-6 mm, suitable for MCAO with reperfusion, or permanent MCAO. Reusable. Pack of 5 filaments.