Learn More
Braintree Scientific Silk Sutures, Non-absorbable, 6-0 100 yds
SDP
Catalog No. NC9201232
Encompass
Silk Sutures, Non-absorbable Basic Sutures for Small Animal Surgery Non-absorbable Silk - The natural softness of silk has been preserved in this high-grade natural suture. This Non-absorbable Suture is composed of a protein called fibroin. Coating: Wax, Characteristics: Silk suture has a smooth flow through tissue while maintaining knot security. Indications: Dekantel silk suture is indicated for use in general soft tissue approximation and/or ligation, including use in cardiovascular, ophthalmic, and neurological procedures. Sizes: USP 5 through 7-0. Construction: Braided. Colors: Black (colorant: logwood extract). Non-sterile 100 yard spool. **Sold for research purposes only and not for human use**
Catalog No. NC9201232 Supplier Braintree Scientific Supplier No. SUTS104
 Restricted Item
Add to Cart
Add to Cart
Silk Sutures, Non-absorbable Basic Sutures for Small Animal Surgery Non-absorbable Silk - The natural softness of silk has been preserved in this high-grade natural suture. This Non-absorbable Suture is composed of a protein called fibroin. Coating: Wax, Characteristics: Silk suture has a smooth flow through tissue while maintaining knot security. Indications: Dekantel silk suture is indicated for use in general soft tissue approximation and/or ligation, including use in cardiovascular, ophthalmic, and neurological procedures. Sizes: USP 5 through 7-0. Construction: Braided. Colors: Black (colorant: logwood extract). Non-sterile 100 yard spool. **Sold for research purposes only and not for human use**