Learn More
Electron Microscopy Sciences Syringe With Tips 6 ml, Autoclavable
SDP
Catalog No. 50949601
Encompass_Preferred
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 6 ml syringe dispenses 100 to 500 uL.
Type: Autoclavable
Catalog No. 50-949-601 Supplier Electron Microscopy Sciences Supplier No. 72484
Used with our repetitive syringe dispenser for dispensing multiple doses of a set volume. The 6 ml syringe dispenses 100 to 500 uL.
Type: Autoclavable