Learn More
Alkali Scientific ASI™ 6.0mm Ceria Stabilized Zirconium Beads, 50/pack
SDP
Catalog No. 503028205
Encompass_Preferred
NEW 6.0mm Ceria Stabilized Zirconium Beads, 50/pack   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.
Catalog No. 50-302-8205 Supplier Alkali Scientific Supplier No. RS70135
NEW 6.0mm Ceria Stabilized Zirconium Beads, 50/pack   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.