Learn More
Med Vet International 6-1/2" (170mm) Angled Rongeur with 4mm Bite, J0224C
Catalog No. 5000585866
Encompass_Preferred
Slightly curved head useful for a wide range of orthopedic procedures including bone spurs on a total femoral head and neck excision.
Catalog No. 50-005-85866 Supplier Med Vet International Supplier No. JOR224C
Slightly curved head useful for a wide range of orthopedic procedures including bone spurs on a total femoral head and neck excision.