Learn More
Sklar Instruments Scissors, Merit Operating Scissors, Sklar 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094103 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Merit Operating Scissors, Sklar, 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Merit Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.
Catalog No. 50-109-4103 Supplier Sklar Instruments Supplier No. 97284
Scissors, Merit Operating Scissors, Sklar, 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Merit Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures.