Learn More
Sklar Instruments Scissors, Surgi-OR Operating Scissors, Sklar 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501094057 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Surgi-OR Operating Scissors, Sklar, 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Surgi-OR Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Catalog No. 50-109-4057 Supplier Sklar Instruments Supplier No. 95284
Scissors, Surgi-OR Operating Scissors, Sklar, 6-1/2 in., Sharp/Sharp, Straight, Smooth, Stainless Steel, EA. Surgi-OR Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.