Learn More
Medchemexpress LLC 6-(1,3-dioxoisoindolin-2-yl)hexanoic acid | 4443-26-9 | MFCD00233499 | 98.0% | 261.27 g·mol⁻¹ | C14H15NO4 | 250 MG
SDP
Catalog No. 5000302470
Encompass_Preferred
O-Phthalimide-C5-acid (6-(1,3-dioxoisoindolin-2-yl)hexanoic acid) is a phthalimide-modified hexanoic acid hapten intended for research use. It features a five-carbon spacer terminating in a carboxyl group, enabling efficient conjugation to carrier proteins for antigen design and immunogen preparation. Supplied with documentation to support quality and safe handling.
  • Terminal carboxyl group for coupling to free amine groups.
  • Suitable as a hapten for antigen design and immunogen preparation.
  • High purity for reproducible conjugation results.
  • Available in multiple pack sizes to support different research scales.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-003-02470 Supplier Medchemexpress LLC Supplier No. 250MGHYW028210
O-Phthalimide-C5-acid (6-(1,3-dioxoisoindolin-2-yl)hexanoic acid) is a phthalimide-modified hexanoic acid hapten intended for research use. It features a five-carbon spacer terminating in a carboxyl group, enabling efficient conjugation to carrier proteins for antigen design and immunogen preparation. Supplied with documentation to support quality and safe handling.
  • Terminal carboxyl group for coupling to free amine groups.
  • Suitable as a hapten for antigen design and immunogen preparation.
  • High purity for reproducible conjugation results.
  • Available in multiple pack sizes to support different research scales.
  • Certificate of analysis and safety data sheet available.