Learn More
Sklar Instruments Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar 6-1/4 in., Blunt, Straight, Horizontally Serrated, Stainless Steel, Disposable. CS of 50.
SDP
Catalog No. 501094259 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar, 6-1/4 in., Blunt,Straight, Horizontally Serrated, Stainless Steel, Disposable. CS of 50. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
Catalog No. 50-109-4259 Supplier Sklar Instruments Supplier No. 962544
Single Use, Econo-Sterile, Rochester-Pean Forceps, Sklar, 6-1/4 in., Blunt,Straight, Horizontally Serrated, Stainless Steel, Disposable. CS of 50. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.