Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-(1-NH2Et)-4-(3-(dimethylamino)Pr)-2-Et-2H-benzo[b]-1,4-oxazin-3(4H)-one diHCl

Catalog No. OTV000412 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000412 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000412 Supplier Sigma Aldrich Supplier No. OTV000412250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant for synthesis of:
  • A chemotype that inhibits botulinum neurotoxin serotype a light chain, p. falciparum malaria, and ebola filovirus
  • Membrane targeting antibiotics
  • 5-ht3a receptor antagonists
  • Histamine h3 antagonists
  • Berberine derivatives for stabilization of g-quadruplex DNA and down regulation of oncogene c-mycreactant for incorporation of basic side chains into cryptolepine scaffold for antimalarial activity

Specifications

Molecular Formula C17H29Cl2N3O2
Linear Formula C17H29Cl2N3O2
MDL Number MFCD21090489
Quantity 5 g
Molecular Weight (g/mol) 378.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.