Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,10-Dibenzyl-N,N'-dimethyl-N,N,N',N'-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]decane-(2R,3R)- 97.0+%, TCI America™
SDP

Catalog No. D3475200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3475200MG 200 mg
D34751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3475200MG Supplier TCI America Supplier No. D3475200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2010983-27-2
Molecular Formula C58H70B2F8N2O2
Synonym (R,R)-TaDiAS-2nd

Specifications

Color White-Yellow
Quantity 200 mg
UN Number 1759
Formula Weight 1000.82
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 6,10-Dibenzyl-N,N′-dimethyl-N,N,N′,N′-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.