Learn More
Ace Glass, Inc. Mantle, 6-10L, for 222.2mm diameter cylindrical flask, 194mm ID x 56.4mm height, 180w-115v, CSA rated
SDP
Catalog No. 50977331
Encompass
6-10L aluminum housed, table-model mantle for 222.2mm diameter cylindrical flask. 194mm I.D. x 56.4mm height. Maximum internal operating temperature is 450 Deg. centigrade. 180w-115v, cULus rated. Power cord included.
Catalog No. 50-977-331 Supplier Ace Glass, Inc. Supplier No. 1205340
6-10L aluminum housed, table-model mantle for 222.2mm diameter cylindrical flask. 194mm I.D. x 56.4mm height. Maximum internal operating temperature is 450 Deg. centigrade. 180w-115v, cULus rated. Power cord included.