Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid 97.0+%, TCI America™
SDP

Catalog No. D42421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42421G 1 g
D42425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42421G Supplier TCI America Supplier No. D42421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55453-87-7
Molecular Formula C16H12O4
Molecular Weight (g/mol) 268.268
MDL Number MFCD00242952
InChI Key QFGMXJOBTNZHEL-UHFFFAOYSA-N
Synonym 6H-11-Oxodibenzo[b,e]oxepin-2-acetic Acid, Isoxepac
PubChem CID 41448
IUPAC Name 2-(11-oxo-6H-benzo[c][1]benzoxepin-2-yl)acetic acid
SMILES C1C2=CC=CC=C2C(=O)C3=C(O1)C=CC(=C3)CC(=O)O

Specifications

Melting Point 142°C
Quantity 1 g
UN Number 2811
Formula Weight 268.27
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,11-Dihydro-11-oxodibenzo[b,e]oxepin-2-acetic Acid
RTECSNumber HQ4110000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.