Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (6-(1H-Pyrazol-1-yl)pyridin-3-yl)methanol

Catalog No. CDS010004 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS010004 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS010004 Supplier Sigma Aldrich Supplier No. CDS01000450MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methyl-1-pentyn-3-ol acts as initiator during the synthesis of propargyl-terminated polylactide by bulk ring-opening polymerization.3-methyl-1-pentyn-3-ol is propargyl alcohol that can be used as:
  • A reactant to synthesize α-methylene cyclic carbonates by reacting with carbon dioxide
  • A reactant in the synthesis of 2,6,9-trisubstituted purine based cdk inhibitors
  • An initiator in the synthesis of polylactide bearing terminal propargyl group via ring-opening polymerization of
  • L-lactide.3-methyl-1-pentyn-3-ol is a sedative and induces hepatic p4503a in mouse

Specifications

Molecular Formula C9H9N3O
Linear Formula C9H9N3O
MDL Number MFCD06658507
Quantity 100 mL
Molecular Weight (g/mol) 175.19
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.