Learn More
Medchemexpress LLC 6-[2-(2,4-difluorophenoxy)-5-(ethylsulfonylmethyl)-3-pyridinyl]-8-methyl-2H-pyrrolo[1,2-a]pyrazin-1-on | 2411226-02-1 | 99.9% | 459.47 g·mol⁻¹ | C22H19F2N3O4S | 5 MG
SDP
Catalog No. 5000262234
Encompass_Preferred
BET bromodomain inhibitor 1 is a selective, orally active inhibitor of bromodomain and extra-terminal (BET) proteins with potent activity against BRD4 (IC50 = 2.6 nM). Supplied as a white to off-white solid with high purity, it is used in epigenetic target validation and anticancer mechanism research.
  • Potent BRD4 inhibition (IC50 2.6 nM).
  • Low-nanomolar binding to BET family bromodomains.
  • High purity suitable for research (99.9%).
  • Soluble in DMSO at 100 mg/mL with warming and sonication.
  • Stable when stored as a powder at -20°C for long-term storage.
Catalog No. 50-002-62234 Supplier Medchemexpress LLC Supplier No. HY1310615MG
BET bromodomain inhibitor 1 is a selective, orally active inhibitor of bromodomain and extra-terminal (BET) proteins with potent activity against BRD4 (IC50 = 2.6 nM). Supplied as a white to off-white solid with high purity, it is used in epigenetic target validation and anticancer mechanism research.
  • Potent BRD4 inhibition (IC50 2.6 nM).
  • Low-nanomolar binding to BET family bromodomains.
  • High purity suitable for research (99.9%).
  • Soluble in DMSO at 100 mg/mL with warming and sonication.
  • Stable when stored as a powder at -20°C for long-term storage.