Learn More
Medchemexpress LLC 6-[2-(2-methyl-1-propan-2-yl-3,4-dihydro-1H-isoquinolin-7-yl)cyclopropyl]naphthalene-2-carboximidamide | 371217-32-2 | 99.3% | 1 ML
SDP
Catalog No. 5000230345
Encompass_Preferred
A-317567 is a small-molecule inhibitor of acid-sensing ion channel 3 (ASIC-3) with a reported IC50 of approximately 1.025 μM. It is supplied for research use as a 10 mM solution (1 ML) in DMSO and as solid aliquots for laboratory studies.
  • Potent ASIC-3 inhibitor (IC50 ≈ 1.025 μM).
  • Available as 10 mM solution (1 ML) in DMSO and as various solid mass sizes.
  • High purity suitable for research (reported purity 99.34%).
  • Molecular formula C27H31N3, molecular weight ≈ 397.6 g/mol.
  • Intended for in vitro and in vivo neuroscience and pain research applications.
Catalog No. 50-002-30345 Supplier Medchemexpress LLC Supplier No. HY12213510MM 1ML
A-317567 is a small-molecule inhibitor of acid-sensing ion channel 3 (ASIC-3) with a reported IC50 of approximately 1.025 μM. It is supplied for research use as a 10 mM solution (1 ML) in DMSO and as solid aliquots for laboratory studies.
  • Potent ASIC-3 inhibitor (IC50 ≈ 1.025 μM).
  • Available as 10 mM solution (1 ML) in DMSO and as various solid mass sizes.
  • High purity suitable for research (reported purity 99.34%).
  • Molecular formula C27H31N3, molecular weight ≈ 397.6 g/mol.
  • Intended for in vitro and in vivo neuroscience and pain research applications.