Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-[2-[4-[(4-Fluorophenyl)phenylmethylene]-1-piperidinyl]ethyl]-7-methyl-5H-thiazolo[3,2-a]pyrimidin-5-on | 93076-89-2 | MFCD00055114 | 99.8% | 459.58 g/mol | C27H26FN3OS | 1 ML
SDP

Catalog No. 5000240048
Encompass_Preferred

R 59-022 is a small-molecule diacylglycerol kinase (DGK) inhibitor that elevates diacylglycerol-dependent protein kinase C activity and acts as a 5-HT receptor antagonist. It is supplied with high reported purity and is commonly used in biochemical and cell-signaling research to investigate DAG/PKC-mediated pathways and receptor pharmacology.

  • High purity suitable for research applications.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid for custom formulation.
  • Molecular weight 459.58 g/mol and chemical formula C27H26FN3OS.
  • Effective probe for studying DGK and PKC signaling pathways.
  • Soluble in common organic solvents, enabling diverse assay formats.

Catalog No. 50-002-40048 Supplier Medchemexpress LLC Supplier No. HY10761310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

R 59-022 is a small-molecule diacylglycerol kinase (DGK) inhibitor that elevates diacylglycerol-dependent protein kinase C activity and acts as a 5-HT receptor antagonist. It is supplied with high reported purity and is commonly used in biochemical and cell-signaling research to investigate DAG/PKC-mediated pathways and receptor pharmacology.

  • High purity suitable for research applications.
  • Provided as a 10 mM solution in DMSO for immediate use.
  • Also available as a solid for custom formulation.
  • Molecular weight 459.58 g/mol and chemical formula C27H26FN3OS.
  • Effective probe for studying DGK and PKC signaling pathways.
  • Soluble in common organic solvents, enabling diverse assay formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.