Learn More
Medchemexpress LLC 6-[2-[4-[(4-fluorophenyl)phenylmethylene]-1-piperidinyl]ethyl]-7-methyl-5H-thiazolo[3,2-a]pyrimidin-5-one | 93076-89-2 | MFCD00055114 | 99.8% | 459.58 g·mol⁻¹ | C27H26FN3OS | 100 MG
SDP
Catalog No. 5000241481
Encompass_Preferred
R 59-022 is a small-molecule diacylglycerol kinase (DGK) inhibitor used in biochemical and cell-based studies to probe DAG/PKC signaling and lipid metabolism. It also functions as a 5-HT receptor antagonist and activates protein kinase C, potentiating thrombin-induced diacylglycerol production in platelets.
  • Inhibits diacylglycerol kinase activity (IC50 ~2.8 μM).
  • Acts as a 5-HT receptor antagonist and activates protein kinase C.
  • Modulates diacylglycerol and phosphatidic acid metabolism in cells.
  • High purity suitable for biochemical assays and cell studies.
  • Offered in solid and solution formats for flexible experimental use.
Catalog No. 50-002-41481 Supplier Medchemexpress LLC Supplier No. HY107613100MG
R 59-022 is a small-molecule diacylglycerol kinase (DGK) inhibitor used in biochemical and cell-based studies to probe DAG/PKC signaling and lipid metabolism. It also functions as a 5-HT receptor antagonist and activates protein kinase C, potentiating thrombin-induced diacylglycerol production in platelets.
  • Inhibits diacylglycerol kinase activity (IC50 ~2.8 μM).
  • Acts as a 5-HT receptor antagonist and activates protein kinase C.
  • Modulates diacylglycerol and phosphatidic acid metabolism in cells.
  • High purity suitable for biochemical assays and cell studies.
  • Offered in solid and solution formats for flexible experimental use.