Learn More
Medchemexpress LLC 6-(2-(4-((4-fluorophenyl)phenylmethylidene)piperidin-1-yl)ethyl)-7-methyl-thiazolo[3,2-a]pyrimidin-5-one | 93076-89-2 | MFCD00055114 | ≥98.0% | 459.58 g/mol | C27H26FN3OS | 25 MG
SDP
Catalog No. 5000240435
Encompass_Preferred
R 59-022 is a small-molecule research reagent that inhibits diacylglycerol kinase (DGK) and functions as a 5-HT receptor antagonist. It is used to modulate protein kinase C (PKC) activation and diacylglycerol-dependent signaling in biochemical and cell-based studies. Chemical identifiers include CAS 93076-89-2, formula C27H26FN3OS, and molecular weight ≈459.58 g/mol.
  • Potent DGK inhibitor (reported IC50 ≈ 2.8 μM).
  • Modulates PKC activation and diacylglycerol signaling.
  • Used in platelet aggregation and cellular signaling studies.
  • High reported purity (≥98%).
  • Provided in small research quantities suitable for assay development.
Catalog No. 50-002-40435 Supplier Medchemexpress LLC Supplier No. HY10761325MG
R 59-022 is a small-molecule research reagent that inhibits diacylglycerol kinase (DGK) and functions as a 5-HT receptor antagonist. It is used to modulate protein kinase C (PKC) activation and diacylglycerol-dependent signaling in biochemical and cell-based studies. Chemical identifiers include CAS 93076-89-2, formula C27H26FN3OS, and molecular weight ≈459.58 g/mol.
  • Potent DGK inhibitor (reported IC50 ≈ 2.8 μM).
  • Modulates PKC activation and diacylglycerol signaling.
  • Used in platelet aggregation and cellular signaling studies.
  • High reported purity (≥98%).
  • Provided in small research quantities suitable for assay development.