Learn More
Medchemexpress LLC 6-[2-(4-aminophenyl)ethenyl]-2-pyridinecarboxylic acid | 115453-99-1 | MFCD05999206 | 98.0% | 240.26 g/mol | C14H12N2O2 | 5 MG
SDP
Catalog No. 5000264922
Encompass_Preferred
CB-7921220 is a small-molecule adenylate cyclase inhibitor (CAS 115453-99-1) intended for laboratory research to modulate cAMP signaling. It shows some isoform preference for adenylate cyclase 1 but limited selectivity versus AC6. The compound is provided as a solid and in ready-to-use DMSO solutions, has reported purity of 98%, and a molecular weight of 240.26 g/mol. For research use only; not for human or clinical applications.
  • Adenylate cyclase inhibitor for cAMP pathway studies.
  • Isoform preference for AC1 with limited selectivity versus AC6.
  • Offered as a solid and in DMSO solution for experimental flexibility.
  • High purity (98.0%) suitable for in vitro assays.
  • Soluble in DMSO; recommended formulation guidance available.
  • Documentation includes datasheet, COA, and SDS.
Catalog No. 50-002-64922 Supplier Medchemexpress LLC Supplier No. HY1018625MG
CB-7921220 is a small-molecule adenylate cyclase inhibitor (CAS 115453-99-1) intended for laboratory research to modulate cAMP signaling. It shows some isoform preference for adenylate cyclase 1 but limited selectivity versus AC6. The compound is provided as a solid and in ready-to-use DMSO solutions, has reported purity of 98%, and a molecular weight of 240.26 g/mol. For research use only; not for human or clinical applications.
  • Adenylate cyclase inhibitor for cAMP pathway studies.
  • Isoform preference for AC1 with limited selectivity versus AC6.
  • Offered as a solid and in DMSO solution for experimental flexibility.
  • High purity (98.0%) suitable for in vitro assays.
  • Soluble in DMSO; recommended formulation guidance available.
  • Documentation includes datasheet, COA, and SDS.