Learn More
Medchemexpress LLC 6-(2,4-Dinitrophenyl)aminohexanoic acid, succinimidyl ester | 82321-04-8 | 98.47% | 394.34 | 100 MG
SDP
Catalog No. 5000428472
Encompass_Preferred
DNP-X, SE (6-(2,4-Dinitrophenyl)aminohexanoic acid, succinimidyl ester) is the DNP-X acid modified by succinimidyl ester. It is an amine-reactive building block for developing a probe, which can be recognized by anti-DNP antibodies. It is also an excellent amine-reactive FRET quencher when paired with Trp or Tyr.
  • Amine-reactive building block
  • Excellent amine-reactive FRET quencher paired with Trp or Tyr
Catalog No. 50-004-28472 Supplier Medchemexpress LLC Supplier No. HYD1050100MG
DNP-X, SE (6-(2,4-Dinitrophenyl)aminohexanoic acid, succinimidyl ester) is the DNP-X acid modified by succinimidyl ester. It is an amine-reactive building block for developing a probe, which can be recognized by anti-DNP antibodies. It is also an excellent amine-reactive FRET quencher when paired with Trp or Tyr.
  • Amine-reactive building block
  • Excellent amine-reactive FRET quencher paired with Trp or Tyr