Learn More
Cayman Chemical 6 2 4-TrImethoxyflavon 10mg
SDP
Catalog No. 502950152
Encompass_Preferred
A flavonoid; inhibits benzo[a]pyrene-induced AhR-dependent transcription in a reporter assay at 2 and 10 µM; decreases LPS-induced production of TNF-α in THP-1 cells (IC50 = 47.7 UG/ml); inhibits cell migration and invasion of HN-30 HNSCC cells at 10 µM
Catalog No. 50-295-0152 Supplier Cayman Chemical Supplier No. 3315010 MG
A flavonoid; inhibits benzo[a]pyrene-induced AhR-dependent transcription in a reporter assay at 2 and 10 µM; decreases LPS-induced production of TNF-α in THP-1 cells (IC50 = 47.7 UG/ml); inhibits cell migration and invasion of HN-30 HNSCC cells at 10 µM