Learn More
Cayman Chemical 6,2',4'-Trimethoxyflavone, Purity: ≥95%, CAS: 720675-74-1, 50 mg
SDP
Catalog No. 502946215
Encompass_Preferred
A flavonoid; inhibits benzo[a]pyrene-induced AhR-dependent transcription in a reporter assay at 2 and 10 µM; decreases LPS-induced production of TNF-α in THP-1 cells (IC50 = 47.7 µg/ml); inhibits cell migration and invasion of HN-30 HNSCC cells at 10 µM
Catalog No. 50-294-6215 Supplier Cayman Chemical Supplier No. 3315050 MG
A flavonoid; inhibits benzo[a]pyrene-induced AhR-dependent transcription in a reporter assay at 2 and 10 µM; decreases LPS-induced production of TNF-α in THP-1 cells (IC50 = 47.7 µg/ml); inhibits cell migration and invasion of HN-30 HNSCC cells at 10 µM