Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(2-(9H-carbazol-9-yl)ethyl)-2,2-dimethyl-2H-chromene-8-carbaldehyde | 1564249-38-2 | 98.0% | 381.47 g·mol^-1 | C26H23NO2 | 1 ML
SDP

Catalog No. 5000227598
Encompass_Preferred

A potent, selective PKCδ inhibitor supplied as a concentrated research solution for laboratory biochemical and cell-based studies.

  • Potent, selective PKCδ inhibitor with reported IC50 ≈ 0.05 μM.
  • Supplied as a 10 mM solution in DMSO, typical vial volume 1 mL.
  • Molecular formula C26H23NO2 and molecular weight 381.47 g·mol⁻1.
  • High purity (reported ≈98%), suitable for research applications.
  • Store in solvent at -80°C for long-term stability or -20°C for short-term storage.
  • For research use only; not intended for human or clinical use.

Catalog No. 50-002-27598 Supplier Medchemexpress LLC Supplier No. HY11780010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A potent, selective PKCδ inhibitor supplied as a concentrated research solution for laboratory biochemical and cell-based studies.

  • Potent, selective PKCδ inhibitor with reported IC50 ≈ 0.05 μM.
  • Supplied as a 10 mM solution in DMSO, typical vial volume 1 mL.
  • Molecular formula C26H23NO2 and molecular weight 381.47 g·mol⁻1.
  • High purity (reported ≈98%), suitable for research applications.
  • Store in solvent at -80°C for long-term stability or -20°C for short-term storage.
  • For research use only; not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.