Learn More
eMolecules​ 6-(2-BROMO-ACETYL)-3,4-DIHYDRO-1H-QUINOLIN-2-ONE | AstaTech | 70639-82-6 | MFCD00173687 | 268.110 | C11H10BrNO2 | 95.000 | BrCC(=O)c1ccc2NC(=O)CCc2c1 | 1g | 112532641
Supplier:  eMolecules​ 112532641
Encompass_Preferred
6-(2-BROMO-ACETYL)-3,4-DIHYDRO-1H-QUINOLIN-2-ONE | AstaTech | 70639-82-6 | MFCD00173687 | 268.110 | C11H10BrNO2 | 95.000 | BrCC(=O)c1ccc2NC(=O)CCc2c1 | 1g | 112532641
Catalog No. NC2229506