Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-(2-Chloroacetyl)-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one

Catalog No. OTV0009461G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0009461G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0009461G Supplier Sigma Aldrich Supplier No. OTV0009461G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-phenyl-1-propanol enantiomers (e-pp) forms the inclusion complexes of cyclic decapeptide (cdp) which were studied using the density functional theory (dft) b3lyp method
  • It was also used to determine the mass transfer kinetics effect on the elution profiles of the 1-phenyl-1-propanol (pp) enantiomers on chiracel ob (cellulose tribenzoate coated on silica gel)

Specifications

CAS 114566-43-7
Molecular Formula C11H10ClNO3
Linear Formula C11H10ClNO3
MDL Number MFCD06763005
Quantity 50 g
Molecular Weight (g/mol) 239.66
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.