Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6:2 Cl-pfaes | 73606-19-6 | MFCD11975311 | >=98.0% | 570.67 g/mol | C8ClF16KO4S | 50 MG
SDP

Catalog No. 5000284921
Encompass_Preferred

6:2 Cl-PFAES (F-53B) is a chlorinated perfluorinated ether sulfonate supplied as the potassium salt and intended for research use in environmental and toxicology studies. Analytical documentation and safety data are provided by the supplier.

  • Potassium salt form suitable for analytical use.
  • Molecular formula C8ClF16KO4S and molecular weight 570.67 g/mol.
  • Purity ≥98.0% by supplier specification.
  • Available in small research pack sizes (50 mg).
  • Documentation available: SDS, COA, and datasheet.
  • Used as a reference standard in PFAS environmental and toxicology studies.
  • Hazard: reproductive toxicity reported; follow SDS handling precautions.

Catalog No. 50-002-84921 Supplier Medchemexpress LLC Supplier No. HYW58374950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6:2 Cl-PFAES (F-53B) is a chlorinated perfluorinated ether sulfonate supplied as the potassium salt and intended for research use in environmental and toxicology studies. Analytical documentation and safety data are provided by the supplier.

  • Potassium salt form suitable for analytical use.
  • Molecular formula C8ClF16KO4S and molecular weight 570.67 g/mol.
  • Purity ≥98.0% by supplier specification.
  • Available in small research pack sizes (50 mg).
  • Documentation available: SDS, COA, and datasheet.
  • Used as a reference standard in PFAS environmental and toxicology studies.
  • Hazard: reproductive toxicity reported; follow SDS handling precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.