Learn More
Medchemexpress LLC HY-N6628 50mg Medchemexpress, 6,2'-Dihydroxyflavone CAS:92439-20-8 Purity:>98%
SDP
Catalog No. 501933962
Encompass_Preferred
Medchemexpress, HY-N6628 50mg 6,2'-Dihydroxyflavone CAS:92439-20-8 6,2'-Dihydroxyflavone is a novel antagonist of GABA A receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3962 Supplier Medchemexpress LLC Supplier No. HYN662850MG
Medchemexpress, HY-N6628 50mg 6,2'-Dihydroxyflavone CAS:92439-20-8 6,2'-Dihydroxyflavone is a novel antagonist of GABA A receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.