Learn More
Medchemexpress LLC 6-((2-fluoro-2-hydroxyethoxy)methyl)-2-(4-(methylamino)phenyl)quinoline | 1374107-54-6 | 98.5% | 326.36 g/mol | C19H19FN2O2 | 25 MG
SDP
Catalog No. 5000213169
Encompass_Preferred
THK-5117 is an arylquinoline research compound that binds tau fibrils and is used in preclinical studies and the development of PET imaging probes for tau pathology. It is supplied as a high-purity solid for laboratory research.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • High purity of 98.5% suitable for analytical and preclinical work.
  • Off-white to light yellow solid for easy handling.
  • Stable when stored under nitrogen at 4°C; in solution store at -80°C for up to six months.
  • Applicable for tau aggregation studies and imaging agent development.
Catalog No. 50-002-13169 Supplier Medchemexpress LLC Supplier No. HY10118225MG
THK-5117 is an arylquinoline research compound that binds tau fibrils and is used in preclinical studies and the development of PET imaging probes for tau pathology. It is supplied as a high-purity solid for laboratory research.
  • High binding affinity to tau fibrils (Ki ≈ 10.5 nM).
  • High purity of 98.5% suitable for analytical and preclinical work.
  • Off-white to light yellow solid for easy handling.
  • Stable when stored under nitrogen at 4°C; in solution store at -80°C for up to six months.
  • Applicable for tau aggregation studies and imaging agent development.