Learn More
Medchemexpress LLC 6-[2-tert-butyl-5-(6-methylpyridin-2-yl)-1H-imidazol-4-yl]quinoxaline | 356559-20-1 | 100.0% | 200 MG
SDP
Catalog No. 5000234166
Encompass_Preferred
SB 525334 is a selective small-molecule inhibitor of transforming growth factor-β receptor I (ALK5) used in preclinical research to block TGF-β signaling. It exhibits potent enzymatic inhibition and is supplied as a solid reagent for laboratory use.
  • selective ALK5 inhibitor with IC50 = 14.3 nM
  • high purity suitable for research use
  • molecular formula C21H21N5 and molecular weight 343.42 g/mol
  • available as a 200 MG solid pack for dosing flexibility
  • stable under recommended storage conditions when stored as directed
Catalog No. 50-002-34166 Supplier Medchemexpress LLC Supplier No. HY12043200MG
SB 525334 is a selective small-molecule inhibitor of transforming growth factor-β receptor I (ALK5) used in preclinical research to block TGF-β signaling. It exhibits potent enzymatic inhibition and is supplied as a solid reagent for laboratory use.
  • selective ALK5 inhibitor with IC50 = 14.3 nM
  • high purity suitable for research use
  • molecular formula C21H21N5 and molecular weight 343.42 g/mol
  • available as a 200 MG solid pack for dosing flexibility
  • stable under recommended storage conditions when stored as directed