Learn More
Ace Glass, Inc. Combo mantle power control, 6 position, 240v 15amp output, percentage timer (pulse) type
SDP
Catalog No. 50977357
Encompass
6-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 240v 15amp output.
Catalog No. 50-977-357 Supplier Ace Glass, Inc. Supplier No. 1206167
6-position Power Control for multi-element combination heating mantle. Power control is the percentage timer (pulse) type. 240v 15amp output.