Learn More
Ace Glass, Inc. Combo mantle power control, 6 position, 240v 15amp output, proportional voltage (steady state) type
SDP
Catalog No. 50977360
Encompass
6-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 240v 15amp output.
Catalog No. 50-977-360 Supplier Ace Glass, Inc. Supplier No. 1206177
6-position Power Control for multi-element combination heating mantle. Power control is the proportional voltage (steady state) type. 240v 15amp output.