Learn More
MSC Baldwin Filters Air Filter Element, Inner, Axial Seal, 1 ea
Catalog No. 5000503691
Encompass
This inner air element is designed for use in engine air filter systems. It features an axial seal design with securely affixed metal end caps and a raised gasket to ensure a proper seal, preventing airborne contaminants from reaching the combustion chamber.
  • Element only
  • Axial seal design
  • Securely affixed metal end caps
  • Raised gasket ensures proper seal
  • Prevents airborne contaminants from reaching combustion chamber
Catalog No. 50-005-03691 Supplier MSC Supplier No. 10937688
This inner air element is designed for use in engine air filter systems. It features an axial seal design with securely affixed metal end caps and a raised gasket to ensure a proper seal, preventing airborne contaminants from reaching the combustion chamber.
  • Element only
  • Axial seal design
  • Securely affixed metal end caps
  • Raised gasket ensures proper seal
  • Prevents airborne contaminants from reaching combustion chamber