Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-(3,4-dibromo-2,5-dioxopyrrol-1-yl)hexanoic acid | 1443214-97-8 | MFCD09813286 | 99.4% | 369.01 g/mol | C10H11Br2NO4 | 50 MG
SDP

Catalog No. 5000204377
Encompass_Preferred

Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide linker used in antibody-drug conjugate synthesis to connect payloads such as MMAF. It is supplied as a white to off-white solid with high purity and is soluble in DMSO.

  • Bifunctional dibromomaleimide linker for ADC synthesis.
  • High purity (99.4%) suitable for research applications.
  • Molecular weight 369.01 g/mol and formula C10H11Br2NO4.
  • Soluble in DMSO at ≥100 mg/mL.
  • Available in small-scale quantities (5-200 mg) for conjugation workflows.
  • Store at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04377 Supplier Medchemexpress LLC Supplier No. HY15708050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibromomaleimide-C5-COOH is a bifunctional dibromomaleimide linker used in antibody-drug conjugate synthesis to connect payloads such as MMAF. It is supplied as a white to off-white solid with high purity and is soluble in DMSO.

  • Bifunctional dibromomaleimide linker for ADC synthesis.
  • High purity (99.4%) suitable for research applications.
  • Molecular weight 369.01 g/mol and formula C10H11Br2NO4.
  • Soluble in DMSO at ≥100 mg/mL.
  • Available in small-scale quantities (5-200 mg) for conjugation workflows.
  • Store at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.