Learn More
Medchemexpress LLC 6-[3,4-dihydroxy-pyrrolid-1-yl]-2-[3-hydroxy-3-methyl-pyrrolid-1-yl]-4-(CF3)pyridine-3-carbonitrile | 2135304-43-5 | 99.1% | 372.34 g·mol⁻1 | C16H19F3N4O3 | 100 MG
SDP
Catalog No. 5000248560
Encompass_Preferred
KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor with a reported IC50 of 0.45 μM, supplied as a high-purity small-molecule research compound suitable for enzymatic, cell-based, and in vivo studies.
  • Potent KHK inhibition with IC50 0.45 μM.
  • High purity, approximately 99.05% by COA.
  • Highly soluble in DMSO (stock solutions up to 250 mg/mL).
  • Formulation guidance provided for in vivo dosing.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years).
  • Datasheet, COA, SDS, and LCMS documentation available for verification.
Catalog No. 50-002-48560 Supplier Medchemexpress LLC Supplier No. HY103478100MG
KHK-IN-2 is a potent and selective ketohexokinase (KHK) inhibitor with a reported IC50 of 0.45 μM, supplied as a high-purity small-molecule research compound suitable for enzymatic, cell-based, and in vivo studies.
  • Potent KHK inhibition with IC50 0.45 μM.
  • High purity, approximately 99.05% by COA.
  • Highly soluble in DMSO (stock solutions up to 250 mg/mL).
  • Formulation guidance provided for in vivo dosing.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years).
  • Datasheet, COA, SDS, and LCMS documentation available for verification.