Learn More
Med Vet International 6 3/4in (17cm) Mayo Supercut Scissors, Straight, J0910S, Each
Catalog No. 5000583986
Encompass_Preferred
These scissors have a specially designed razor-sharp edge that cuts through tissue like butter. The lower blade has micro fine serrations to prevent tissue slippage. Black finger rings for easy identification.
Catalog No. 50-005-83986 Supplier Med Vet International Supplier No. JOR910S
These scissors have a specially designed razor-sharp edge that cuts through tissue like butter. The lower blade has micro fine serrations to prevent tissue slippage. Black finger rings for easy identification.