Learn More
Sklar Instruments Scissors, Surgi-OR Mayo Dissecting Scissors, Sklar 6-3/4 in., Blunt/Blunt, Straight, Serrated, Stainless Steel, EA.
SDP
Catalog No. 501094066 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Surgi-OR Mayo Dissecting Scissors, Sklar, 6-3/4 in., Blunt/Blunt, Straight, Serrated, Stainless Steel, EA. Surgi-OR Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. This product is straight with serrated, blunt/blunt tips and a length of 6-3/4 inches.
Catalog No. 50-109-4066 Supplier Sklar Instruments Supplier No. 95329
Scissors, Surgi-OR Mayo Dissecting Scissors, Sklar, 6-3/4 in., Blunt/Blunt, Straight, Serrated, Stainless Steel, EA. Surgi-OR Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and multiple settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. This product is straight with serrated, blunt/blunt tips and a length of 6-3/4 inches.