Learn More
Medchemexpress LLC 6-(3-hydroxypropyl)-2-(trimethyl-2-oxo-2,3-dihydro-benzimidazol-5-yl)-benzoisoquinoline-1,3-dione | 2080306-23-4 | MFCD30480933 | 99.4% | 429.47 g·mol⁻¹ | C25H23N3O4 | 50 MG
SDP
Catalog No. 5000241017
Encompass_Preferred
BAY-299 is a benzoisoquinolinedione chemical probe originally described by Bayer that potently inhibits the bromodomain-containing proteins BRPF2 (BRD1) and the second bromodomain of TAF1 and TAF1L. It is used in biochemical, structural, and cellular studies to probe bromodomain function and selectivity, with reported biochemical IC50s in the low-nanomolar range and validated activity in cell-based assays.
Catalog No. 50-002-41017 Supplier Medchemexpress LLC Supplier No. HY10742450MG
BAY-299 is a benzoisoquinolinedione chemical probe originally described by Bayer that potently inhibits the bromodomain-containing proteins BRPF2 (BRD1) and the second bromodomain of TAF1 and TAF1L. It is used in biochemical, structural, and cellular studies to probe bromodomain function and selectivity, with reported biochemical IC50s in the low-nanomolar range and validated activity in cell-based assays.