Learn More
Beckman Coulter Sealing Tubes
Catalog No. NC2307878
Encompass_Preferred
The Beckman Coulter Microfuge Tubes are polypropylene tubes with attached domed caps that provide an excellent moisture barrier and secure seal. They are available in 0.5mL and 1.5mL volumes, and come in a variety of colors for easy sample identification. The attached caps eliminate loose lids and prevent contamination. These microcentrifuge tubes have high centrifugal stability up to 15,000 x g RCF. They are ideal for sample storage, transport, and processing in clinical, research, and testing labs. The tubes are certified DNAse, RNAse and Pyrogen free. They are non-cytotoxic and non-hemolytic.
Catalog No. NC2307878 Supplier Beckman Coulter Supplier No. 345830
The Beckman Coulter Microfuge Tubes are polypropylene tubes with attached domed caps that provide an excellent moisture barrier and secure seal. They are available in 0.5mL and 1.5mL volumes, and come in a variety of colors for easy sample identification. The attached caps eliminate loose lids and prevent contamination. These microcentrifuge tubes have high centrifugal stability up to 15,000 x g RCF. They are ideal for sample storage, transport, and processing in clinical, research, and testing labs. The tubes are certified DNAse, RNAse and Pyrogen free. They are non-cytotoxic and non-hemolytic.