Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-[(3S,4S)-4-amino-3-methyl-2-oxa-8-azaspiro[4.5]decan-8-yl]-3-(2,3-dichlorophenyl)-2-methyl-3,4-di | 2411321-29-2 | 99.4% | C20H24Cl2N4O2 | 10MG
SDP

Catalog No. 5000261495
Encompass_Preferred

IACS-15414 is a small-molecule, orally bioavailable inhibitor of the SHP2 phosphatase with a reported IC50 of 122 nM, supplied for preclinical biochemical, cellular, and in vivo research. The compound has a molecular weight of 423.34, formula C20H24Cl2N4O2, and a reported purity of 99.39%.

  • Potent SHP2 inhibitor (IC50 122 nM).
  • Orally bioavailable and suitable for in vivo studies.
  • High reported purity (99.39%).
  • Soluble in DMSO (100 mg/mL) and compatible with published in vivo formulation protocols.
  • Molecular weight 423.34 and formula C20H24Cl2N4O2.
  • Recommended storage as powder at -20°C or 4°C for shorter term stability.
  • Available in small research pack sizes for preclinical use.

Catalog No. 50-002-61495 Supplier Medchemexpress LLC Supplier No. HY13290110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IACS-15414 is a small-molecule, orally bioavailable inhibitor of the SHP2 phosphatase with a reported IC50 of 122 nM, supplied for preclinical biochemical, cellular, and in vivo research. The compound has a molecular weight of 423.34, formula C20H24Cl2N4O2, and a reported purity of 99.39%.

  • Potent SHP2 inhibitor (IC50 122 nM).
  • Orally bioavailable and suitable for in vivo studies.
  • High reported purity (99.39%).
  • Soluble in DMSO (100 mg/mL) and compatible with published in vivo formulation protocols.
  • Molecular weight 423.34 and formula C20H24Cl2N4O2.
  • Recommended storage as powder at -20°C or 4°C for shorter term stability.
  • Available in small research pack sizes for preclinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.