Learn More
Medchemexpress LLC 6-(4-(1-aminocyclobutyl)phenyl)-5-phenylnicotinamide | 1357158-81-6 | >98.0% | 343.42 g/mol | C22H21N3O | 10 MG
SDP
Catalog No. 5000294889
Encompass_Preferred
AKT-IN-1 is a small-molecule allosteric inhibitor of the AKT kinase family used for pathway and cancer research. It inhibits AKT with an IC50 of 1.042 μM, has CAS 1357158-81-6, and is supplied as a research-grade solid with high purity for biochemical and cellular assays.
  • Allosteric AKT inhibition with IC50 = 1.042 μM.
  • High chemical purity for reproducible results.
  • Suitable for PI3K/Akt/mTOR pathway studies and cancer research.
  • Solid form enabling flexible solution preparation.
  • Molecular weight 343.42 g/mol and formula C22H21N3O for accurate dosing.
  • Intended for research use only.
Catalog No. 50-002-94889 Supplier Medchemexpress LLC Supplier No. HY1829610MG
AKT-IN-1 is a small-molecule allosteric inhibitor of the AKT kinase family used for pathway and cancer research. It inhibits AKT with an IC50 of 1.042 μM, has CAS 1357158-81-6, and is supplied as a research-grade solid with high purity for biochemical and cellular assays.
  • Allosteric AKT inhibition with IC50 = 1.042 μM.
  • High chemical purity for reproducible results.
  • Suitable for PI3K/Akt/mTOR pathway studies and cancer research.
  • Solid form enabling flexible solution preparation.
  • Molecular weight 343.42 g/mol and formula C22H21N3O for accurate dosing.
  • Intended for research use only.