Learn More
Medchemexpress LLC 6-(4-(4-methylthiazol-2-yl)-1H-imidazol-5-yl)benzo[d]thiazole hydrochloride | 2459963-17-6 | MFCD32633583 | 99.2% | 334.85 g/mol | C14H11ClN4S2 | 10 MG
SDP
Catalog No. 5000308312
Encompass_Preferred
TP0427736 hydrochloride is a research-grade small-molecule ALK5 (TGF-β receptor I) inhibitor supplied as the hydrochloride salt. The compound is provided as a high-purity solid intended for in vitro biochemical and cellular studies, with documented solubility in water and DMSO and recommended cold storage for long-term stability.
  • High purity suitable for research assays.
  • Hydrochloride salt form improves aqueous and DMSO solubility.
  • Documented solubility values aid experimental planning.
  • Stable when stored as powder at recommended cold temperatures.
  • Appropriate for pathway, target validation, and pharmacology studies.
Catalog No. 50-003-08312 Supplier Medchemexpress LLC Supplier No. HY118528A10MG
TP0427736 hydrochloride is a research-grade small-molecule ALK5 (TGF-β receptor I) inhibitor supplied as the hydrochloride salt. The compound is provided as a high-purity solid intended for in vitro biochemical and cellular studies, with documented solubility in water and DMSO and recommended cold storage for long-term stability.
  • High purity suitable for research assays.
  • Hydrochloride salt form improves aqueous and DMSO solubility.
  • Documented solubility values aid experimental planning.
  • Stable when stored as powder at recommended cold temperatures.
  • Appropriate for pathway, target validation, and pharmacology studies.