Learn More
Medchemexpress LLC 6-(4-(7-(tert-butyl)-5-(4-chloro-3-fluorophenyl)...piperazin-1-yl) nicotinic acid | 2014368-93-3 | MFCD18251438 | 99.4% | 565.04 | C30H30ClFN4O4 | 10 MG
SDP
Catalog No. 5000287268
Encompass_Preferred
I-287 is an orally active, selective protease-activated receptor 2 (PAR2) inhibitor used in research to modulate Gαq and Gα12/13-mediated signaling. It is supplied as a high-purity small-molecule compound characterized for pharmacology studies in cellular and animal models.
  • Selective PAR2 inhibitor.
  • Orally active small molecule.
  • Acts as a negative allosteric regulator of Gαq and Gα12/13.
  • Characterized for modulation of downstream signaling pathways.
  • High purity (~99.4%) suitable for research applications.
  • Available in research-scale quantities, such as 10 mg.
Catalog No. 50-002-87268 Supplier Medchemexpress LLC Supplier No. HY14801610MG
I-287 is an orally active, selective protease-activated receptor 2 (PAR2) inhibitor used in research to modulate Gαq and Gα12/13-mediated signaling. It is supplied as a high-purity small-molecule compound characterized for pharmacology studies in cellular and animal models.
  • Selective PAR2 inhibitor.
  • Orally active small molecule.
  • Acts as a negative allosteric regulator of Gαq and Gα12/13.
  • Characterized for modulation of downstream signaling pathways.
  • High purity (~99.4%) suitable for research applications.
  • Available in research-scale quantities, such as 10 mg.