Learn More
AdipoGen 6-4-Acetamido-1 8-naphthalami
SDP
Catalog No. 502561853
Encompass_Preferred
Chemical. CAS 172227-59-7. Formula C20H20N2O5. MW 368.38. Synthetic. The highly fluorescent label 4-acetylamino-1,8-naphthalimide-N-caproic acid has two fused benzene rings containing a closed anhydride ring and therefore is a flat and planar molecule which can easily intercalate or act as a groove binder with DNA/RNA sequences. For the study of in situ hydridization with complementary synthetic oligonucleotides ODNs, tagging with non-radioactive labels has gained importance, not only for medical diagnostics or clinical genetics, but also detection and isolation of DNA for research purposes. The pentamethylene chain in 6 acts as a linker molecule and keeps the target at a reasonable distance to avoid direct interaction with oligonucleotides.
Catalog No. 50-256-1853 Supplier AdipoGen Supplier No. CDXA0038M050
Chemical. CAS 172227-59-7. Formula C20H20N2O5. MW 368.38. Synthetic. The highly fluorescent label 4-acetylamino-1,8-naphthalimide-N-caproic acid has two fused benzene rings containing a closed anhydride ring and therefore is a flat and planar molecule which can easily intercalate or act as a groove binder with DNA/RNA sequences. For the study of in situ hydridization with complementary synthetic oligonucleotides ODNs, tagging with non-radioactive labels has gained importance, not only for medical diagnostics or clinical genetics, but also detection and isolation of DNA for research purposes. The pentamethylene chain in 6 acts as a linker molecule and keeps the target at a reasonable distance to avoid direct interaction with oligonucleotides.