Learn More
Medchemexpress LLC 6-[(4-bromo-2-thienyl)methoxy]-9H-purin-2-amine | 192441-08-0 | MFCD23703756 | >=98.0% | 326.17 g·mol⁻¹ | C10H8BrN5OS | 5 MG
SDP
Catalog No. 5000221913
Encompass_Preferred
Lomeguatrib is a potent O6-methylguanine-DNA methyltransferase (MGMT) inhibitor used in research to sensitize cells to alkylating agents; it shows low-nanomolar activity in biochemical and cell assays.
  • Potent MGMT inhibitor (IC50 9 nM cell-free; ~6 nM in MCF-7 cells).
  • Used to sensitize cells to alkylating agents in research applications.
  • Available in small research pack sizes: 1 mg, 5 mg, 10 mg.
  • Molecular weight 326.17 g·mol⁻¹; chemical formula C10H8BrN5OS.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 2 years).
  • Supplied as a research-grade small molecule for in vitro studies.
Catalog No. 50-002-21913 Supplier Medchemexpress LLC Supplier No. HY136685MG
Lomeguatrib is a potent O6-methylguanine-DNA methyltransferase (MGMT) inhibitor used in research to sensitize cells to alkylating agents; it shows low-nanomolar activity in biochemical and cell assays.
  • Potent MGMT inhibitor (IC50 9 nM cell-free; ~6 nM in MCF-7 cells).
  • Used to sensitize cells to alkylating agents in research applications.
  • Available in small research pack sizes: 1 mg, 5 mg, 10 mg.
  • Molecular weight 326.17 g·mol⁻¹; chemical formula C10H8BrN5OS.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 2 years).
  • Supplied as a research-grade small molecule for in vitro studies.