Learn More
Medchemexpress LLC 6-(4-chlorophenyl)-2,3-dihydro-N-[(1S)-2-hydroxy-1-methylethyl]-2-(1-methyl-1H-pyrazol-4-yl)-3-oxo-4 | 2242464-44-2 | MFCD32693912 | >98.0% | C18H18ClN5O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13582910MG
Encompass_Preferred
BAY 2416964 is a small-molecule aryl hydrocarbon receptor (AhR) antagonist developed as a clinical-stage oncology candidate. It inhibits AhR signaling in vitro, has shown cellular potency in reported assays, and has been evaluated in early-phase clinical studies as an immunomodulatory agent to enhance anti-tumor immune responses.
  • Small-molecule aryl hydrocarbon receptor antagonist.
  • Demonstrated cellular potency, reported IC50 ~341 nM.
  • Investigated in early-phase clinical studies for solid tumors.
  • High chemical purity typical for supplied samples (>98.0%).
  • Suitable for preclinical pharmacology and mechanism-of-action studies.
  • Molecular formula C18H18ClN5O3; molecular weight ~387.8.
Catalog No. 50-002-58766