Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-[[4-(cyclopropylmethyl)-1-piperazinyl]methyl]-2-(5-fluoro-indol-4-yl)-4-(4-morpholinyl)-thieno-pyrimidine | 955977-50-1 | 99.0% | 50 MG
SDP

Catalog No. 5000230705
Encompass_Preferred

PI-3065 is a selective small-molecule inhibitor of PI3K p110δ used for research applications. It potently inhibits p110δ (IC50 5 nM; Ki 1.5 nM) and shows substantially lower activity against other PI3K isoforms, and is supplied as a solid for in vitro and in vivo studies.

  • Potent p110δ inhibition (IC50 5 nM; Ki 1.5 nM).
  • Reduced activity against p110α (IC50 910 nM), p110β (IC50 600 nM), p110γ (>10,000 nM).
  • Purity approximately 98.97%.
  • Chemical formula C27H31FN6OS; molecular weight 506.64 g/mol.
  • Appearance: white to off-white solid.
  • Soluble in DMSO (25 mg/mL) and formulatable for in vivo dosing.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small-mass pack sizes (for example 50 mg).

Catalog No. 50-002-30705 Supplier Medchemexpress LLC Supplier No. HY1223550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PI-3065 is a selective small-molecule inhibitor of PI3K p110δ used for research applications. It potently inhibits p110δ (IC50 5 nM; Ki 1.5 nM) and shows substantially lower activity against other PI3K isoforms, and is supplied as a solid for in vitro and in vivo studies.

  • Potent p110δ inhibition (IC50 5 nM; Ki 1.5 nM).
  • Reduced activity against p110α (IC50 910 nM), p110β (IC50 600 nM), p110γ (>10,000 nM).
  • Purity approximately 98.97%.
  • Chemical formula C27H31FN6OS; molecular weight 506.64 g/mol.
  • Appearance: white to off-white solid.
  • Soluble in DMSO (25 mg/mL) and formulatable for in vivo dosing.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small-mass pack sizes (for example 50 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.