Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-((4-hydroxybutyl)amino)hexyl 2-hexyldecanoate | 2644752-88-3 | 98.0% | 427.70 | C26H53NO3 | 100 MG
SDP

Catalog No. 5000280739
Encompass_Preferred

6-((4-hydroxybutyl)amino)hexyl 2-hexyldecanoate is a synthetic lipid reagent for research use, commonly employed in the formulation of nanomaterials and lipid-based nucleic acid delivery systems. It is supplied as a purified compound intended for laboratory applications.

  • Purity 98.0%.
  • Molecular formula C26H53NO3; molecular weight 427.70.
  • CAS number 2644752-88-3.
  • Available in 25 mg, 50 mg, 100 mg, and larger quantities by quote.
  • Suitable for synthesis of nanomaterials and lipid nanoparticle formulation.
  • Storage: pure form -20°C for long-term, 4°C for medium-term; in solvent -80°C (6 months), -20°C (1 month).
  • Appearance colorless to light yellow liquid.

Catalog No. 50-002-80739 Supplier Medchemexpress LLC Supplier No. HYW440815100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-((4-hydroxybutyl)amino)hexyl 2-hexyldecanoate is a synthetic lipid reagent for research use, commonly employed in the formulation of nanomaterials and lipid-based nucleic acid delivery systems. It is supplied as a purified compound intended for laboratory applications.

  • Purity 98.0%.
  • Molecular formula C26H53NO3; molecular weight 427.70.
  • CAS number 2644752-88-3.
  • Available in 25 mg, 50 mg, 100 mg, and larger quantities by quote.
  • Suitable for synthesis of nanomaterials and lipid nanoparticle formulation.
  • Storage: pure form -20°C for long-term, 4°C for medium-term; in solvent -80°C (6 months), -20°C (1 month).
  • Appearance colorless to light yellow liquid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.