Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-((4-hydroxybutyl)amino)hexyl 2-hexyldecanoate | 2644752-88-3 | 98.0% | 427.70 | C26H53NO3 | 25 MG
SDP

Catalog No. 5000281148
Encompass_Preferred

6-((4-Hydroxybutyl)amino)hexyl 2-hexyldecanoate is a synthetic lipid for research applications, commonly used to synthesize nanomaterials and as a component in lipid-based nucleic acid delivery systems. Supplied as a liquid at high purity, it is intended for formulation development and delivery studies that require reproducible material quality.

  • High purity: 98.0%.
  • Molecular formula C26H53NO3; molecular weight 427.70.
  • Appearance: colorless to light yellow liquid.
  • Storage: pure form -20°C for 3 years, 4°C for 2 years.
  • Intended use: for research use only.

Catalog No. 50-002-81148 Supplier Medchemexpress LLC Supplier No. HYW44081525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-((4-Hydroxybutyl)amino)hexyl 2-hexyldecanoate is a synthetic lipid for research applications, commonly used to synthesize nanomaterials and as a component in lipid-based nucleic acid delivery systems. Supplied as a liquid at high purity, it is intended for formulation development and delivery studies that require reproducible material quality.

  • High purity: 98.0%.
  • Molecular formula C26H53NO3; molecular weight 427.70.
  • Appearance: colorless to light yellow liquid.
  • Storage: pure form -20°C for 3 years, 4°C for 2 years.
  • Intended use: for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.